PIPES
Glass bottle for solids, 250 g
Catalog # ROTH9156.3
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Piperazine-1,4-bis(2-ethanesulphonic acid), 2,2'-(Piperazine-1,4-diyl)diethanesulphonic acid, Piperazine-N,N'-bis-2-ethanesulphonic acid, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid
- CAS Number:5625-37-6
- Formula:C₈H₁₈N₂O₆S₂
- MDL Number:MFCD00006159
- Molecular Weight:302.37 g/mol
- Merck Index:12,07633
- Boiling Pt:499 °C (1013 hPa)
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:227-057-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



