n-Undecane
Glass bottle, 100 mL
Catalog # ROTH8844.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Undecane
- CAS Number:1120-21-4
- Formula:H₃C(CH₂)₉CH₃
- MDL Number:MFCD00008959
- Molecular Weight:156.31 g/mol
- Boiling Pt:196 °C (1013 hPa)
- Melting Pt:–26.7 °C
- Density:0.7416 g/cm³ (20 °C)
- Flash Pt:60 °C
- Storage Temperature:Ambient
- EINECS:214-300-6
- UN:2330
- ADR:3,III
Specifications
Specifications
- Pack type:Glass bottle
- Pk:100 mL



