Order Entry
Export
Orders LinkContactUsLinkComponent
 

Acetonitrile

Glass bottle, 2,5 L
Catalog # ROTH8824.2
Flammable
Irritant
Supplier:   Roth Carl
Acetonitrile
Catalog # ROTH8824.2
Supplier:  Roth Carl
CAS Number:  75-05-8
Acetonitrile
Acetonitrile

Glass bottle, 2,5 L

CAS Number:  75-05-8
Price
156,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
  • CAS Number:75-05-8
  • Formula:H₃CCN
  • MDL Number:MFCD00001878
  • Molecular Weight:41.05 g/mol
  • Merck Index:13,00071
  • Boiling Pt:81.6 °C (1013 hPa)
  • Melting Pt:–45.7 °C
  • Density:0.782 g/cm³ (20 °C)
  • Flash Pt:2 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-835-2
  • UN:1648
  • ADR:3,II
  • REACH:01-2119471307-38
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications