1,4-Dithioerythritol
Glass bottle for solids, 25 g
Catalog # ROTH8814.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol
- CAS Number:6892-68-8
- Formula:C₄H₁₀O₂S₂
- MDL Number:MFCD00063750
- Molecular Weight:154.25 g/mol
- Boiling Pt:286 °C (1013 hPa)
- Melting Pt:82…83 °C
- Storage Temperature:Ambient
- EINECS:229-998-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



