Order Entry
Export
Orders LinkContactUsLinkComponent
1,4-Dithioerythritol

1,4-Dithioerythritol

Glass bottle for solids, 5 g
Catalog # ROTH8814.1
Irritant
Supplier:   Roth Carl
1,4-Dithioerythritol
Catalog # ROTH8814.1
Supplier:  Roth Carl
CAS Number:  6892-68-8
1,4-Dithioerythritol

Glass bottle for solids, 5 g

CAS Number:  6892-68-8
Price
90,90 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol
  • CAS Number:6892-68-8
  • Formula:C₄H₁₀O₂S₂
  • MDL Number:MFCD00063750
  • Molecular Weight:154.25 g/mol
  • Boiling Pt:286 °C (1013 hPa)
  • Melting Pt:82…83 °C
  • Storage Temperature:Ambient
  • EINECS:229-998-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications