Order Entry
Export
Orders LinkContactUsLinkComponent
n-Pentane

n-Pentane

Glass bottle, 2,5 L
Catalog # ROTH8720.3
Flammable
Health
Irritant
Environment
Supplier:   Roth Carl
n-Pentane
Catalog # ROTH8720.3
Supplier:  Roth Carl
CAS Number:  109-66-0
n-Pentane
n-Pentane

Glass bottle, 2,5 L

CAS Number:  109-66-0
Price
95,40 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Pentane
  • CAS Number:109-66-0
  • Formula:H₃C(CH₂)₃CH₃
  • MDL Number:MFCD00009498
  • Molecular Weight:72.15 g/mol
  • Merck Index:13,07193
  • Boiling Pt:35…36 °C (1013 hPa)
  • Melting Pt:–130 °C
  • Density:0.63 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • Storage Temperature:Ambient
  • EINECS:203-692-4
  • UN:1265
  • ADR:3,II
  • REACH:01-2119459286-30
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications