Order Entry
Export
Orders LinkContactUsLinkComponent
n-Heptane

n-Heptane

Glass bottle, 1 L
Catalog # ROTH8654.1
Flammable
Health
Irritant
Environment
Supplier:   Roth Carl
n-Heptane
Catalog # ROTH8654.1
Supplier:  Roth Carl
CAS Number:  142-82-5
n-Heptane
n-Heptane

Glass bottle, 1 L

CAS Number:  142-82-5
Price
44,60 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  high purity solvents
  • CAS Number:142-82-5
  • Formula:H₃C(CH₂)₅CH₃
  • MDL Number:MFCD00009544
  • Molecular Weight:100.20 g/mol
  • Merck Index:13,04679
  • Boiling Pt:98.4 °C (1013 hPa)
  • Melting Pt:–90.6 °C
  • Density:0.6838 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • EINECS:205-563-8
  • UN:1206
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications