Order Entry
Export
Orders LinkContactUsLinkComponent
Soda lime

Soda lime

Glass bottle, 1 L
Catalog # ROTH8652.1
Corrosive
Supplier:   Roth Carl
Soda lime
Catalog # ROTH8652.1
Supplier:  Roth Carl
CAS Number:  8006-28-8
Soda lime

Glass bottle, 1 L

CAS Number:  8006-28-8
Price
51,80 €
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:8006-28-8
  • MDL Number:MFCD00134094
  • Merck Index:13,08640
  • UN:1907
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications