Order Entry
Export
Orders LinkContactUsLinkComponent
Cesium chloride

Cesium chloride

Glass bottle for solids, 1 kg
Catalog # ROTH8627.2
Supplier:   Roth Carl
Cesium chloride
Catalog # ROTH8627.2
Supplier:  Roth Carl
CAS Number:  7647-17-8
Cesium chloride

Glass bottle for solids, 1 kg

CAS Number:  7647-17-8
Price
965,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Cesium Chloride
  • CAS Number:7647-17-8
  • Formula:CsCl
  • MDL Number:MFCD00010955
  • Molecular Weight:168.36 g/mol
  • Merck Index:13,02021
  • Boiling Pt:1303 °C (1013 hPa)
  • Melting Pt:646 °C
  • Density:3.7808 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-600-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications