tri-Calcium phosphate
Glass bottle for solids, 250 g
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Calcium phosphate, Tricalcium orthophosphate, penta-Calcium hydroxide triphosphate, Calcium phosphate tribasic
- CAS Number:7758-87-4
- Formula:Ca₃(PO₄)₂
- MDL Number:MFCD00015984
- Molecular Weight:310.18 g/mol
- Merck Index:12,01741
- Density:3.14 g/cm³ (20 °C)
- EINECS:231-840-8
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



