Tray, Rotilabo®
520×325×90 mm
Catalog # ROTH8105.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Dishes, made of stainless steel, chrome-nickel plated.
- With folding handles on two sides
- Very good temperature resistance
- Easy to clean
- Autoclavable
Suitable for food applications.
Specifications
- W×D×H:520×325×90 mm