Potassium hydroxide
25 kg
Catalog # ROTH7986.5
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Potash caustic, Caustic potash
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Merck Index:12,07806
- Boiling Pt:1320 °C (1013 hPa)
- Melting Pt:360 °C
- Density:2.04 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-181-3
- UN:1813
- ADR:8,II
- REACH:01-2119487136-33
Specifications
Specifications
- Pk:25 kg



