Potassium hexacyanoferrate(III)
Glass bottle for solids, 1 kg
ROTH7971.1
: Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide
- :13746-66-2
- :K₃Fe(CN)₆
- :MFCD00011392
- :329.25 g/mol
- :13,07712
- :1.85 g/cm³ (20 °C)
- :Ambient
- :237-323-3
- Pack type:Glass bottle for solids
- Pk:1 kg