Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium hydroxide 50% in aqueous solution

Potassium hydroxide 50% in aqueous solution

10 L
Catalog # ROTH7949.2
Corrosive
Supplier:   Roth Carl
Potassium hydroxide 50% in aqueous solution
Catalog # ROTH7949.2
Supplier:  Roth Carl
CAS Number:  1310-58-3
Potassium hydroxide 50% in aqueous solution

10 L

CAS Number:  1310-58-3
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Potash caustic, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.11 g/mol
  • Storage Temperature:Ambient
  • EINECS:215-181-3
  • UN:1814
  • ADR:8,II

Documents

Specifications

Specifications

  • Pk:
    10 L

Specifications