Potassium hydroxide 50% in aqueous solution
Glass bottle, 1 L
Catalog # ROTH7949.1
Supplier: Roth Carl
Potassium hydroxide 50% in aqueous solution
Catalog # ROTH7949.1
Supplier: Roth Carl
CAS Number: 1310-58-3
Jump to:
Product Details & Documents
Synonyms: Potash caustic, Caustic potash
- CAS Number:1310-58-3
- Formula:KOH
- MDL Number:MFCD00003553
- Molecular Weight:56.11 g/mol
- Storage Temperature:Ambient
- EINECS:215-181-3
- UN:1814
- ADR:8,II
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



