Acrylamide, 4x crystallised
Glass bottle for solids, 100 g
ROTH7906.1
: Prop-2-enamide
- :79-06-1
- :C₃H₅NO
- :MFCD00008032
- :71.08 g/mol
- :232 °C (1013 hPa)
- :84.5 °C
- :1.122 g/cm³ (20 °C)
- :138 °C
- :Refrigerator
- :201-173-7
- :2074
- :6.1,III
- Pack type:Glass bottle for solids
- Pk:100 g