Cesium chloride
Glass bottle for solids, 100 g
Catalog # ROTH7878.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Cesium Chloride
- CAS Number:7647-17-8
- Formula:CsCl
- MDL Number:MFCD00010955
- Molecular Weight:168.36 g/mol
- Merck Index:13,02021
- Boiling Pt:1303 °C (1013 hPa)
- Melting Pt:646 °C
- Density:3.7808 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-600-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



