Order Entry
Export
Orders LinkContactUsLinkComponent
Acrylamide, 2x crystallised

Acrylamide, 2x crystallised

Glass bottle for solids, 500 g
Catalog # ROTH7871.1
Toxic
Health
Supplier:   Roth Carl
Acrylamide, 2x crystallised
Catalog # ROTH7871.1
Supplier:  Roth Carl
CAS Number:  79-06-1
Acrylamide, 2x crystallised
Acrylamide, 2x crystallised

Glass bottle for solids, 500 g

CAS Number:  79-06-1
Price
64,30 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Boiling Pt:232 °C (1013 hPa)
  • Melting Pt:84.5 °C
  • Density:1.122 g/cm³ (20 °C)
  • Flash Pt:138 °C
  • Storage Temperature:Refrigerator
  • EINECS:201-173-7
  • UN:2074
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications