Order Entry
Export
Orders LinkContactUsLinkComponent
Ammonium acetate

Ammonium acetate

Glass bottle for solids, 2,5 kg
Catalog # ROTH7869.3
Irritant
Supplier:   Roth Carl
Ammonium acetate
Catalog # ROTH7869.3
Supplier:  Roth Carl
CAS Number:  631-61-8
Ammonium acetate
Ammonium acetate

Glass bottle for solids, 2,5 kg

CAS Number:  631-61-8
Price
Inquire For Price
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Acetic acid ammonium salt
  • CAS Number:631-61-8
  • Formula:H₃CCOONH₄
  • MDL Number:MFCD00013066
  • Molecular Weight:77.08 g/mol
  • Merck Index:13,00495
  • Melting Pt:114 °C
  • Density:1.17 g/cm³ (20 °C)
  • Flash Pt:136 °C
  • Storage Temperature:Ambient
  • EINECS:211-162-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2,5 kg

Specifications