Triethylamine
Glass bottle, 1 L
Catalog # ROTHX875.3
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: (Diethylamino)ethane, TEA
- CAS Number:121-44-8
- Formula:(C₂H₅)₃N
- MDL Number:MFCD00009051
- Molecular Weight:101.19 g/mol
- Merck Index:13,09740
- Boiling Pt:90 °C (1013 hPa)
- Melting Pt:–115 °C
- Density:0.729 g/cm³ (20 °C)
- Flash Pt:–11 °C
- Storage Temperature:Ambient
- EINECS:204-469-4
- UN:1296
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



