Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-(Dimethylamino)benzaldehyde

Glass bottle for solids, 100 g
Catalog # ROTHX867.2
Supplier:   Roth Carl
4-(Dimethylamino)benzaldehyde
Catalog # ROTHX867.2
Supplier:  Roth Carl
CAS Number:  100-10-7
4-(Dimethylamino)benzaldehyde
4-(Dimethylamino)benzaldehyde

Glass bottle for solids, 100 g

CAS Number:  100-10-7
Price
145,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
  • CAS Number:100-10-7
  • Formula:(CH₃)₂NC₆H₄CHO
  • MDL Number:MFCD00003381
  • Molecular Weight:149.19 g/mol
  • Merck Index:13,03257
  • Boiling Pt:176…177 °C (1013 hPa)
  • Melting Pt:72 °C
  • Density:1.0254 g/cm³ (99.9 °C)
  • Flash Pt:>100 °C
  • Storage Temperature:Ambient
  • EINECS:202-819-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications