4-(Dimethylamino)benzaldehyde
Glass bottle for solids, 25 g
ROTHX867.1
: Ehrlich's Reagent, 4-(N,N-Dimethylamino)benzaldehyde, p-Dimethylamino-Benzaldehyde, p-Dimethylaminobenzaldehyde, p-(Dimethylamino)benzaldehyde
- :100-10-7
- :(CH₃)₂NC₆H₄CHO
- :MFCD00003381
- :149.19 g/mol
- :13,03257
- :176…177 °C (1013 hPa)
- :72 °C
- :1.0254 g/cm³ (99.9 °C)
- :>100 °C
- :Ambient
- :202-819-0
- Pack type:Glass bottle for solids
- Pk:25 g