Order Entry
Export
Orders LinkContactUsLinkComponent
Charcoal activated

Charcoal activated

Glass bottle for solids, 500 g
Catalog # ROTHX865.2
Supplier:   Roth Carl
Charcoal activated
Catalog # ROTHX865.2
Supplier:  Roth Carl
CAS Number:  7440-44-0
Charcoal activated

Glass bottle for solids, 500 g

CAS Number:  7440-44-0
Price
53,90 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Carbon activated, Activated carbon, Activated charcoal
  • CAS Number:7440-44-0
  • Formula:C
  • MDL Number:MFCD00133992
  • Molecular Weight:12.01 g/mol
  • Merck Index:13,01818
  • Melting Pt:3550 °C
  • Density:2 g/cm³ (20 °C)
  • EINECS:231-153-3
  • UN:1362
  • ADR:4.2,III
  • REACH:01-2119488894-16

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications