0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Charcoal activated
Charcoal activated
Charcoal activated
Glass bottle for solids, 250 g
ROTHX865.1
:
Roth Carl
Charcoal activated
ROTHX865.1
:
Roth Carl
:
7440-44-0
Charcoal activated
Glass bottle for solids, 250 g
:
7440-44-0
34,20 €
1 * 250 g
-
+
:
Carbon activated, Activated carbon, Activated charcoal
:
7440-44-0
:
C
:
MFCD00133992
:
12.01 g/mol
:
13,01818
:
3550 °C
:
2 g/cm³ (20 °C)
:
231-153-3
:
1362
:
4.2,III
:
01-2119488894-16
Pack type:
Glass bottle for solids
Pk:
250 g