Order Entry
Export
Orders LinkContactUsLinkComponent
Hydrochloric acid 32%

Hydrochloric acid 32%

Glass bottle, 2,5 L
Catalog # ROTHP074.2
Corrosive
Irritant
Supplier:   Roth Carl
Hydrochloric acid 32%
Catalog # ROTHP074.2
Supplier:  Roth Carl
CAS Number:  7647-01-0
Hydrochloric acid 32%

Glass bottle, 2,5 L

CAS Number:  7647-01-0
Price
46,60 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Boiling Pt:110 °C (1013 hPa)
  • Melting Pt:–30 °C
  • Density:1.18 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications