Order Entry
Export
Orders LinkContactUsLinkComponent
Butyl acetate

Butyl acetate

Glass bottle, 2,5 L
Catalog # ROTHP036.2
Flammable
Irritant
Supplier:   Roth Carl
Butyl acetate
Catalog # ROTHP036.2
Supplier:  Roth Carl
CAS Number:  123-86-4
Butyl acetate
Butyl acetate

Glass bottle, 2,5 L

CAS Number:  123-86-4
Price
172,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Butyl ethanoate, Acetic acid butyl ester, Acetic acid n-butyl ester, n-Butyl acetate
  • CAS Number:123-86-4
  • Formula:CH₃COO(CH₂)₃CH₃
  • MDL Number:MFCD00009445
  • Molecular Weight:116.16 g/mol
  • Merck Index:13,01534
  • Boiling Pt:126 °C (1013 hPa)
  • Melting Pt:–76 °C
  • Density:0.875 g/cm³ (20 °C)
  • Flash Pt:25 °C
  • Storage Temperature:Ambient
  • EINECS:204-658-1
  • UN:1123
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications