Sodium chloride
5 kg
Catalog # ROTHP029.3
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Saline, NaCl, Sea salt, Halite
- CAS Number:7647-14-5
- Formula:NaCl
- MDL Number:MFCD00003477
- Molecular Weight:58.44 g/mol
- Merck Index:14,08599
- Boiling Pt:1413 °C (1013 hPa)
- Melting Pt:801 °C
- Density:2.16 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-598-3
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:5 kg



