Caffeine
Glass bottle for solids, 50 g
Catalog # ROTHN815.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine
- CAS Number:58-08-2
- Formula:C₈H₁₀N₄O₂
- MDL Number:MFCD00005758
- Molecular Weight:194.19 g/mol
- Merck Index:13,01636
- Melting Pt:235…239 °C
- Density:1.23 g/cm³ (19 °C)
- Storage Temperature:Ambient
- EINECS:200-362-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



