Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ammonium acetate

25 kg
Catalog # ROTHT872.5
Irritant
Supplier:   Roth Carl
Ammonium acetate
Catalog # ROTHT872.5
Supplier:  Roth Carl
CAS Number:  631-61-8
Ammonium acetate
Ammonium acetate

25 kg

CAS Number:  631-61-8
Price
691,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  Acetic acid ammonium salt
  • CAS Number:631-61-8
  • Formula:H₃CCOONH₄
  • MDL Number:MFCD00013066
  • Molecular Weight:77.08 g/mol
  • Merck Index:13,00495
  • Melting Pt:114 °C
  • Density:1.17 g/cm³ (20 °C)
  • Flash Pt:136 °C
  • Storage Temperature:Ambient
  • EINECS:211-162-9
Specifications

Specifications

  • Pk:
    25 kg

Specifications