Ammonium acetate
Glass bottle for solids, 500 g
Catalog # ROTHT872.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: Acetic acid ammonium salt
- CAS Number:631-61-8
- Formula:H₃CCOONH₄
- MDL Number:MFCD00013066
- Molecular Weight:77.08 g/mol
- Merck Index:13,00495
- Melting Pt:114 °C
- Density:1.17 g/cm³ (20 °C)
- Flash Pt:136 °C
- Storage Temperature:Ambient
- EINECS:211-162-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



