Order Entry
Export
Orders LinkContactUsLinkComponent
May Grunwald's eosin-methylene blue solution

May Grunwald's eosin-methylene blue solution

Glass bottle, 1 L
Catalog # ROTHT863.2
Flammable
Toxic
Health
Supplier:   Roth Carl
May Grunwald's eosin-methylene blue solution
Catalog # ROTHT863.2
Supplier:  Roth Carl
CAS Number:  
May Grunwald's eosin-methylene blue solution

Glass bottle, 1 L

CAS Number:  
Price
45,60 €
Log in for availability
1 * 1 L
Product Details & Documents
  • MDL Number:MFCD00131580
  • Storage Temperature:Ambient
  • UN:1230
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications