Order Entry
Export
Orders LinkContactUsLinkComponent
Lugol's iodine solution

Lugol's iodine solution

Glass bottle, 1 L
Catalog # ROTHN052.2
Supplier:   Roth Carl
Lugol's iodine solution
Catalog # ROTHN052.2
Supplier:  Roth Carl
CAS Number:  
Lugol's iodine solution

Glass bottle, 1 L

CAS Number:  
Price
51,80 €
Log in for availability
1 * 1 L
Product Details & Documents
  • Density:1.01 g/cm³ (20 °C)
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications