Order Entry
Export
Orders LinkContactUsLinkComponent
L(-)-Tyrosine

L(-)-Tyrosine

Glass bottle for solids, 500 g
Catalog # ROTHT207.3
Supplier:   Roth Carl
L(-)-Tyrosine
Catalog # ROTHT207.3
Supplier:  Roth Carl
CAS Number:  60-18-4
L(-)-Tyrosine

Glass bottle for solids, 500 g

CAS Number:  60-18-4
Price
213,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
  • CAS Number:60-18-4
  • Formula:C₉H₁₁NO₃
  • MDL Number:MFCD00002606
  • Molecular Weight:181.19 g/mol
  • Merck Index:13,09907
  • Boiling Pt:351 °C (1013 hPa)
  • Melting Pt:285 °C
  • Density:1.41 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-460-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications