L(-)-Proline
Glass bottle for solids, 25 g
Catalog # ROTHT205.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
- CAS Number:147-85-3
- Formula:C₅H₉NO₂
- MDL Number:MFCD00064318
- Molecular Weight:115.13 g/mol
- Merck Index:13,07871
- Boiling Pt:245 °C (1013 hPa)
- Melting Pt:220…222 °C
- Density:1.35 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:205-702-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



