Sodium hydroxide 32% in aqueous solution
Glass bottle, 1 L
Catalog # ROTHT196.1
Supplier: Roth Carl
Sodium hydroxide 32% in aqueous solution
Catalog # ROTHT196.1
Supplier: Roth Carl
CAS Number: 1310-73-2
Jump to:
Product Details & Documents
Synonyms: Caustic soda
- CAS Number:1310-73-2
- Formula:NaOH
- MDL Number:MFCD00003548
- Molecular Weight:40 g/mol
- Boiling Pt:119…135 °C (1013 hPa)
- Melting Pt:9…10 °C
- Density:1,53 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-185-5
- UN:1824
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L



