n-Heptane
Glass bottle, 2,5 L
Catalog # ROTHT174.2
Supplier: Roth Carl
Jump to:
Product Details & Documents
Synonyms: high purity solvents
- CAS Number:142-82-5
- Formula:H₃C(CH₂)₅CH₃
- MDL Number:MFCD00009544
- Molecular Weight:100.20 g/mol
- Merck Index:13,04679
- Boiling Pt:98.4 °C (1013 hPa)
- Melting Pt:–90.6 °C
- Density:0.6838 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- EINECS:205-563-8
- UN:1206
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



