Order Entry
Export
Orders LinkContactUsLinkComponent
Milk, powder

Milk, powder

Glass bottle for solids, 500 g
Catalog # ROTHT145.2
Supplier:   Roth Carl
Milk, powder
Catalog # ROTHT145.2
Supplier:  Roth Carl
CAS Number:  999999-99-4
Milk, powder

Glass bottle for solids, 500 g

CAS Number:  999999-99-4
Price
28,60 €
Log in for availability
1 * 500 g
Product Details & Documents
  • CAS Number:999999-99-4
  • EINECS:310-127-6

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications