Milk, powder
Glass bottle for solids, 250 g
Catalog # ROTHT145.1
Supplier: Roth Carl
Jump to:
Product Details & Documents
- CAS Number:999999-99-4
- EINECS:310-127-6
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



