Order Entry
Export
Orders LinkContactUsLinkComponent
 

Zinc oxide

1 kg
Catalog # ROTHT114.2
Environment
Supplier:   Roth Carl
Zinc oxide
Catalog # ROTHT114.2
Supplier:  Roth Carl
CAS Number:  1314-13-2
Zinc oxide

1 kg

CAS Number:  1314-13-2
Price
134,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Zinci oxidum, Zinc (II) oxide
  • CAS Number:1314-13-2
  • Formula:ZnO
  • MDL Number:MFCD00011300
  • Molecular Weight:81.39 g/mol
  • Merck Index:13,10200
  • Melting Pt:1975 °C
  • Density:5.61 g/cm³ (25 °C)
  • Storage Temperature:Ambient
  • EINECS:215-222-5
  • UN:3077
  • ADR:9,III
  • REACH:01-2119463881-32
Specifications

Specifications

  • Pk:
    1 kg

Specifications