Order Entry
Export
Orders LinkContactUsLinkComponent
Chloral hydrate

Chloral hydrate

Glass bottle for solids, 500 g
Catalog # ROTHK318.1
Toxic
Supplier:   Roth Carl
Chloral hydrate
Catalog # ROTHK318.1
Supplier:  Roth Carl
CAS Number:  302-17-0
Chloral hydrate
Chloral hydrate

Glass bottle for solids, 500 g

CAS Number:  302-17-0
Price
145,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  2,2,2-Trichloroethane-1,1-diol
  • CAS Number:302-17-0
  • Formula:Cl₃CCH(OH)₂
  • MDL Number:MFCD00044479
  • Molecular Weight:165.4 g/mol
  • Merck Index:12,02113
  • Boiling Pt:97.5 °C (1013 hPa)
  • Melting Pt:52 °C
  • Density:1.901 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:206-117-5
  • ADR:6.1,III
  •  :Density: 1.901 g/cm³ (20 °C)
  • UN:2811
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications