Order Entry
Export
Orders LinkContactUsLinkComponent
Ammonium chloride

Ammonium chloride

Glass bottle for solids, 1 kg
Catalog # ROTHK298.2
Irritant
Supplier:   Roth Carl
Ammonium chloride
Catalog # ROTHK298.2
Supplier:  Roth Carl
CAS Number:  12125-02-9
Ammonium chloride
Ammonium chloride

Glass bottle for solids, 1 kg

CAS Number:  12125-02-9
Price
60,80 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Ammonium muriate, Salmiac
  • CAS Number:12125-02-9
  • Formula:NH₄Cl
  • MDL Number:MFCD00011420
  • Molecular Weight:53.49 g/mol
  • Merck Index:13,00510
  • Melting Pt:335 °C
  • Density:1.5256 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:235-186-4

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications