Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium thiocyanate

Potassium thiocyanate

Glass bottle for solids, 500 g
Catalog # ROTHP753.1
Irritant
Supplier:   Roth Carl
Potassium thiocyanate
Catalog # ROTHP753.1
Supplier:  Roth Carl
CAS Number:  333-20-0
Potassium thiocyanate

Glass bottle for solids, 500 g

CAS Number:  333-20-0
Price
99,20 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Potassium rhodanide
  • CAS Number:333-20-0
  • Formula:KSCN
  • MDL Number:MFCD00011413
  • Molecular Weight:97.18 g/mol
  • Merck Index:12,07866
  • Boiling Pt:500 °C (1013 hPa)
  • Melting Pt:175 °C
  • Density:1.61 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:206-370-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications