Order Entry
Export
Orders LinkContactUsLinkComponent
Potassium hexacyanoferrate(III)

Potassium hexacyanoferrate(III)

Glass bottle for solids, 1 kg
Catalog # ROTHP746.2
Supplier:   Roth Carl
Potassium hexacyanoferrate(III)
Catalog # ROTHP746.2
Supplier:  Roth Carl
CAS Number:  13746-66-2
Potassium hexacyanoferrate(III)

Glass bottle for solids, 1 kg

CAS Number:  13746-66-2
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Potassium ferricyanide(III), Tripotassium hexacyanoferrate, Potassium ferricyanide
  • CAS Number:13746-66-2
  • Formula:K₃Fe(CN)₆
  • MDL Number:MFCD00011392
  • Molecular Weight:329.25 g/mol
  • Merck Index:13,07712
  • Density:1.85 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:237-323-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications