Order Entry
Export
Orders LinkContactUsLinkComponent
Lead(II) acetate trihydrate

Lead(II) acetate trihydrate

Glass bottle for solids, 500 g
Catalog # ROTHP739.1
Health
Environment
Supplier:   Roth Carl
Lead(II) acetate trihydrate
Catalog # ROTHP739.1
Supplier:  Roth Carl
CAS Number:  6080-56-4
Lead(II) acetate trihydrate
Lead(II) acetate trihydrate

Glass bottle for solids, 500 g

CAS Number:  6080-56-4
Price
120,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate
  • CAS Number:6080-56-4
  • Formula:(H₃CCOO)₂Pb·3H₂O
  • MDL Number:MFCD00150023
  • Molecular Weight:379.34 g/mol
  • Merck Index:13,05415
  • Boiling Pt:280 °C (1013 hPa)
  • Melting Pt:75 °C
  • Density:2.55 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:206-104-4
  • UN:1616
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications