Order Entry
Export
Orders LinkContactUsLinkComponent
Diammonium hydrogen orthophosphate

Diammonium hydrogen orthophosphate

Glass bottle for solids, 1 kg
Catalog # ROTHP727.2
Supplier:   Roth Carl
Diammonium hydrogen orthophosphate
Catalog # ROTHP727.2
Supplier:  Roth Carl
CAS Number:  7783-28-0
Diammonium hydrogen orthophosphate

Glass bottle for solids, 1 kg

CAS Number:  7783-28-0
Price
124,00 €
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
  • CAS Number:7783-28-0
  • Formula:(NH₄)₂HPO₄
  • MDL Number:MFCD00010891
  • Molecular Weight:132.06 g/mol
  • Merck Index:13,00546
  • Melting Pt:185 °C
  • Density:1.619 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-987-8
  • REACH:01-2119490974-22
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications