Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Buffer solutionpH 4,00±0,02 (20 °C) 1 * 1 L
ROTHP712.1
:
Roth Carl
Buffer solutionpH 4,00±0,02 (20 °C) 1 * 1 L
ROTHP712.1
:
Roth Carl