0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Potassium hydroxide
Potassium hydroxide 1 mol/l (1 N) in aqueous solution
Potassium hydroxide 1 mol/l (1 N) in aqueous solution
5 L
ROTHK017.2
:
Roth Carl
Potassium hydroxide 1 mol/l (1 N) in aqueous solution
ROTHK017.2
:
Roth Carl
:
1310-58-3
Potassium hydroxide 1 mol/l (1 N) in aqueous solution
5 L
:
1310-58-3
85,10 €
1 * 5 L
-
+
:
Potash caustic, Caustic potash
:
1310-58-3
:
KOH
:
MFCD00003553
:
56.11 g/mol
:
Ambient
:
215-181-3
:
1814
:
8,III
Pk:
5 L