Ethyl acetate
Glass bottle, 1 L
ROTH7338.1
: Acetic acid ethyl ester
- :141-78-6
- :CH₃COOC₂H₅
- :MFCD00009171
- :88.11 g/mol
- :13,03792
- :77.1 °C (1013 hPa)
- :–83 °C
- :0.902 g/cm³ (20 °C)
- :–4 °C
- :Ambient
- :205-500-4
- :1173
- :3,II
- :01-2119475103-46
- Pack type:Glass bottle
- Pk:1 L
- Cat. No.:ROTH7338.1
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health



