Order Entry
Export
Orders LinkContactUsLinkComponent
Sorbic acid

Sorbic acid

Glass bottle for solids, 250 g
Catalog # ROTH7158.1
Irritant
Supplier:   Roth Carl
Sorbic acid
Catalog # ROTH7158.1
Supplier:  Roth Carl
CAS Number:  110-44-1
Sorbic acid
Sorbic acid

Glass bottle for solids, 250 g

CAS Number:  110-44-1
Price
60,10 €
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid
  • CAS Number:110-44-1
  • Formula:CH₃CH=CHCH=CHCOOH
  • MDL Number:MFCD00002703
  • Molecular Weight:112.13 g/mol
  • Merck Index:13,08793
  • Boiling Pt:228 °C (1013 hPa)
  • Melting Pt:132…135 °C
  • Density:1.204 g/cm³ (20 °C)
  • Flash Pt:127 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:203-768-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications