Order Entry
Export
Orders LinkContactUsLinkComponent
Toluene

Toluene

Glass bottle, 1 L
Catalog # ROTH7115.1
Flammable
Health
Irritant
Supplier:   Roth Carl
Toluene
Catalog # ROTH7115.1
Supplier:  Roth Carl
CAS Number:  108-88-3
Toluene
Toluene

Glass bottle, 1 L

CAS Number:  108-88-3
Price
51,40 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Methylbenzene, Phenylmethane, FM approval
  • CAS Number:108-88-3
  • Formula:C₆H₅CH₃
  • MDL Number:MFCD00008512
  • Molecular Weight:92.14 g/mol
  • Merck Index:13,09607
  • Boiling Pt:110.6 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:0.867 g/cm³ (20 °C)
  • Flash Pt:4 °C
  • Storage Temperature:Ambient
  • EINECS:203-625-9
  • UN:1294
  • ADR:3,II
  • REACH:05-2114615130-69
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications